PROTEOSTAT® Protein Refolding and Aggregation Sensing Kit, Enzo Life Sciences
Catalog # 10033-622
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Size:1 Kit
- Cat. No.:10033-622
- Supplier no.:51040-KP002
Specifications
About this item
ProteoStat® Protein refolding and aggregation sensing kit provides a fractional factorial matrix design to facilitate screening of refolding parameters for a specific protein in different buffers, and a simple, homogenous assay format for monitoring protein aggregation using a red-emitting aggregation-sensitive dye.