Native Hsv-1 Antigen
10033-586
:
- Protein/Peptide Type:Native
- Size:200 µl
- Form:Liquid
- Protein/Peptide Name:Hsv-1 Antigen
- Purity:~80%
- Formulation:PBS containing 0.1% Triton X-100
- Cat. No.:10033-586
- Supplier no.:630-126-0200
From herpes simplex virus type 1. Inactivated by 0.1% Triton X-100.