Native C. Trachomatis
Catalog # 10033-558
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/Peptide Type:Native
- Size:1 ml
- Protein/Peptide Name:C. Trachomatis
- Formulation:Liquid, PBS containing 0.05% Triton X-100
- Purification:Cell lysate.
- Cat. No.:10033-558
- Supplier no.:630-124-1000
Specifications
About this item
Cell lysate ideal for use as a broad positive control.
From Chlamydia trachomatis, serotype LGV2 strain 434. Inactivated by heat treatment and 0.05% Triton X-100.