ContactUsLinkComponent
White Silicone Septum with PTFE Bonded Face, Ace Glass
  89077-550
 :  Ace Glass
Small Business Enterprise
undefined
White Silicone Septum with PTFE Bonded Face, Ace Glass
  89077-550
 :  Ace Glass
 :  12913-40
Small Business Enterprise
  • Ø:
    32 mm
  • Description:
    Septum
  • Thickness:
    2 mm
  • Cat. No.:
    89077-550
  • Supplier no.:
    12913-40
Fits GPI 33-430 phenolic caps
PTFE faced silicone resists coring when punctured via syringe needle. For use with the Screw Caps, Black Plastic, by Ace Glass.