ContactUsLinkComponent
Envirocide Disinfectant, Metrex
Envirocide Disinfectant, Metrex
Catalog # 89132-882
Supplier:  Metrex
Envirocide Disinfectant, Metrex
Catalog # 89132-882
Supplier:  Metrex
Supplier Number:  13-3324
Specifications
  • Description:
    Envirocide Spray
  • Size:
    24 oz.
  • Cat. No.:
    89132-882
  • Supplier no.:
    13-3324
Specifications
About this item
Envirocide is a convenient, ready-to-use, intermediate-level surface disinfectant which is effective against TB, HBV, HCV, viruses (hydrophilic and lipophilic), bacteria (including MRSA and VRE) and fungi
It is safe for all areas of the facility including NICU, operating rooms, isolation rooms, patient care areas and laboratories. When used as directed, it will also effectively clean and decontaminate critical and semi-critical instrumentation. Envirocide is safe for use on non-porous surfaces. It is perfect for cleaning and decontamination of environmental and medical device surfaces.