ErgoTec® Safety Scraper, Unger®
89094-543
- Color:Green
- Dimensions:11/2"
- Material:Stainless steel
- Cat. No.:89094-543
- Supplier no.:UNG SR040
ErgoTec® Safety Scraper with rubber grip prevents scraper from slipping in hand.
- Stainless steel safety blades
- Rubber grip prevents scraper from slipping in hand
- Unique locking system prevents blade from sliding out
Push the nonslip trigger forward and blade is ready for use, pushing the blade forward a second time makes blade easy to change.
: Includes one scraper.