ContactUsLinkComponent
Boardwalk® Light Duty Scour Pad
  21899-525
undefined
Boardwalk® Light Duty Scour Pad
  21899-525
 :  BWK198
  • Color:
    White
  • Description:
    Light Duty Scour Pads
  • Dimensions:
    6 x 9"
  • Material:
    Fiber, synthetic
  • Environmentally Preferable:
  • Cat. No.:
    21899-525
  • Supplier no.:
    PAD 198
Ideal for cleaning glass, stainless steel, porcelain, chrome, copper and ceramic surfaces.
Won't scratch or dull most surfaces.
 : Includes 20 pads per carton.