Sulfuric acid ≥96%, Gigabit®
Catalog # 71009-382
Supplier: KMG
CAS Number:
Specifications
- Pack type:Poly bottle, bagged
- Size:1 gal.
- Cat. No.:71009-382
- Supplier no.:460-064105
Specifications
Specification Test Results
- Assay (H2SO4):95.50 - 97.00%
- Assay (H2O):3.00 - 5.00%
- Color:6 APHA
- Appearance:Conforms
- Substance Reducing KMnO4:Conforms
- Chloride (Cl):100 ppb
- Nitrrate (NO3):100 ppb
- Phosphate (PO4):100 ppb
- Aluminum (Al):2 ppb
- Antimony (Sb):1 ppb
- Arsenic (As):1 ppb
- Barium (Ba):1 ppb
- Beryllium (Be):1 ppb
- Bismuth (Bi):1 ppb
- Boron (B):1 ppb
- Cadmium (Cd):1 ppb
- Chromium (Cr):1 ppb
- Cobalt (Co):1 ppb
- Copper (Cu):1 ppb
- Gallium (Ga):1 ppb
- Germanium (Ge):1 ppb
- Gold (Au):1 ppb
- Iron (Fe):5 ppb
- Lead (Pb):1 ppb
- Lithium (Li):1 ppb
- Magnesium (Mg):1 ppb
- Manganese (Mn):1 ppb
- Molybdenum (Mo):1 ppb
- Nickel (Ni):1 ppb
- Niobium (Nb):1 ppb
- Potassium (K):2 ppb
- Silicon (Si):5 ppb
- Silver (Ag):1 ppb
- Sodium (Na):2 ppb
- Strontium (Sr):1 ppb
- Tantalum (Ta):1 ppb
- Thallium (Tl):1 ppb
- Tin (Sn):1 ppb
- Titanium (Ti):1 ppb
- Vanadium (V):1 ppb
- Zinc (Zn):2 ppb
- 0.065μ Particle Count:20000 par/ml
- 0.1μ Particle Count:10000 par/ml
- 0.15μ Particle Count:2000 par/ml
- 0.2μ Particle Count:250 par/ml
- 0.5μ Particle Count:25par/ml
Specification Test Results
About this item
:
:98.08 g/mol
Boiling Pt:∼335 °C (1013 hPa)
Boiling Pt:~ 335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
UN:1830
ADR:8,II
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589