Methyl oleate reference substance for gas chromatography, Supelco®
Catalog # EM1.09743.0005
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle, pre-packed
- Size:5 ml
- Cat. No.:EM1.09743.0005
- Supplier no.:1.09743.0005
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-20 °C
- Molar Mass:296.49 g/mol
- Boiling Point:202 - 210 °C (20 hPa)
- Flash Point:113 °C
- Density:0.87 g/cm3 (20 °C)
Specification Test Results
About this item
:
:296.49 g/mol
CAS Number:112-62-9
:67762-38-3
Formula:C₁₉H₃₆O₂
MDL Number:MFCD00009578