Diiodomethane for separation of minerals mixtures, Supelco®
Catalog # EM1.06053.0100
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:EM1.06053.0100
- Supplier no.:1.06053.0100
Specifications
Specification Test Results
- Solubility:0.8 g/l (25 °C)
- Melting Point:6 °C
- Molar Mass:267.84 g/mol
- Boiling Point:181 °C (1013 hPa) (decomposition)
- Flash Point:>110 °C
- Density:3.3 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:267.84 g/mol
Merck Index:12,06143
Boiling Pt:182 °C (1013 hPa)
Boiling Pt:182 °C (1013 hPa)
Melting Pt:6 °C
Density:3.32537 g/cm³ (20 °C)
Flash Pt:110 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-841-5
UN:2810
ADR:6.1,III
:GHS06
CAS Number:75-11-6
Formula:CH₂I₂
MDL Number:MFCD00001079