Order Entry
Puerto Rico
ContactUsLinkComponent
Ammonium sulfide ≥20% in aqueous solution, GR analytical reagent, Supelco®
Flammable
Corrosive
Ammonium sulfide ≥20% in aqueous solution, GR analytical reagent, Supelco®
Catalog # EM1.05442.1000
Supplier:  MilliporeSigma
CAS Number:  
Flammable
Corrosive
Ammonium sulfide ≥20% in aqueous solution, GR analytical reagent, Supelco®
Catalog # EM1.05442.1000
Supplier:  MilliporeSigma
Supplier Number:  1.05442.1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    EM1.05442.1000
  • Supplier no.:
    1.05442.1000

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Vapor Pressure:
    450 hPa (20 °C)
  • Flash Point:
    15 °C
  • Explosion Limit:
    4 - 46 %(V)
  • Density:
    0.997 g/cm3 (20 °C)
  • pH:
    9.5 (200 g/l, H2O, 25 °C)
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

About this item

 :
 :68.14
EINECS:235-223-4
UN:2683
ADR:8 (3, 6.1),II
 :GHS02
 :GHS05
CAS Number:12135-76-1
Formula:H₈N₂S
MDL Number:MFCD00010892