Dimethyl glyoxime disodium salt octahydrate, GR for analysis, Supelco®
Catalog # EM1.03061.0100
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic bottle
- Size:100 g
- Cat. No.:EM1.03061.0100
- Supplier no.:1.03061.0100
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:304.21 g/mol
- Bulk Density:300 kg/m3
Specification Test Results
About this item
:
Storage Temperature:Ambient
EINECS:262-523-2
CAS Number:75006-64-3
Formula:C₄H₆N₂Na₂O₂·8H₂O
MDL Number:MFCD00150288