Order Entry
Puerto Rico
ContactUsLinkComponent
Quinhydrone ACS, Reag. Ph. Eur., GR for analysis, Supelco®
Quinhydrone ACS, Reag. Ph. Eur., GR for analysis, Supelco®
Catalog # EM1.02284.0100
Supplier:  MilliporeSigma
CAS Number:  
Quinhydrone ACS, Reag. Ph. Eur., GR for analysis, Supelco®
Catalog # EM1.02284.0100
Supplier:  MilliporeSigma
Supplier Number:  1.02284.0100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g
  • Cat. No.:
    EM1.02284.0100
  • Supplier no.:
    1.02284.0100

Specifications

Specification Test Results

  • Solubility:
    4 g/l (20 °C)
  • Melting Point:
    168 - 171 °C
  • Molar Mass:
    218.21 g/mol
  • Bulk Density:
    300 kg/m3
  • Density:
    1.40 g/cm3 (20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

About this item

 :
 :218.21 g/mol
Merck Index:13,08148
Melting Pt:168…171 °C
Melting Pt:168 - 171 °C
Density:1.35 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-387-6
UN:2811
ADR:6.1,III
 :GHS06
 :GHS09
CAS Number:106-34-3
Formula:C₆H₄(OH)₂·C₆H₄O₂
MDL Number:MFCD00010310