Bis(cyclohexanone)oxalyldihydrazone ≥99% (by perchloric acid titration), GR for analysis (reagent for copper), Supelco®
Catalog # EM1.01841.0025
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:25 g
- Cat. No.:EM1.01841.0025
- Supplier no.:1.01841.0025
Specifications
Specification Test Results
- Assay (perchloric acid titration):≥ 99 %
- Identity (IR-spectrum):conforms
- Melting range (lower value):≥ 208 °C
- Melting range (upper value):≤ 214 °C
- Suitability for copper determiN/Ation:conforms
Specification Test Results
About this item
:
:278.36 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:370-81-0
Formula:C₁₄H₂₂N₄O₂
MDL Number:MFCD00001659