CurTec Packo Container, White PP, Qorpak®
: Qorpak
This High Performance Packaging line provides the highest level of product protection and added value through quality, functionality, ease of use, and design
- Water tight closure sold separately
- Open and close with just a quarter-turn
- Tamper evident feature
- Smooth inside
- UN marking for solids
Suggested for Pharmaceuticals, Fine Chemicals, Food Ingredients and Hazardous Waste. Prevents moisture contamination and is airtight.
Applications include shipping and stocking valuable fine chemicals, API’s and intermediates as well as finished products in powder, tablet and paste format. The line is backed by a quality assurance program which includes HACCP, ISO certification and a Drug Master File for all packaging products.