Order Entry
Puerto Rico
ContactUsLinkComponent
4-(2-Chloroethyl)morpholine hydrochloride for synthesis, Sigma-Aldrich®
Toxic
Corrosive
4-(2-Chloroethyl)morpholine hydrochloride for synthesis, Sigma-Aldrich®
Catalog # EM8.41417.0100
Supplier:  MilliporeSigma
CAS Number:  
Toxic
Corrosive
4-(2-Chloroethyl)morpholine hydrochloride for synthesis, Sigma-Aldrich®
Catalog # EM8.41417.0100
Supplier:  MilliporeSigma
Supplier Number:  8.41417.0100
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    100 g
  • Cat. No.:
    EM8.41417.0100
  • Supplier no.:
    8.41417.0100

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    182 - 184 °C (decomposition)
  • Molar Mass:
    186.08 g/mol
  • pH:
    4.5 - 5 (10 g/l, H2O, 20 °C)
  • Ignition Point:
    365 °C

Specification Test Results

About this item

 :
 :186.08 g/mol
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ200
EINECS:222-881-2
UN:2923
 :GHS06
 :GHS05
CAS Number:3647-69-6
Formula:C₆H₁₃Cl₂NO
MDL Number:MFCD00012797