Order Entry
Puerto Rico
ContactUsLinkComponent
9-Fluorenone for synthesis, Sigma-Aldrich®
Irritant
Environment
9-Fluorenone for synthesis, Sigma-Aldrich®
Catalog # EM8.03977.0250
Supplier:  MilliporeSigma
CAS Number:  
Irritant
Environment
9-Fluorenone for synthesis, Sigma-Aldrich®
Catalog # EM8.03977.0250
Supplier:  MilliporeSigma
Supplier Number:  8.03977.0250
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g
  • Cat. No.:
    EM8.03977.0250
  • Supplier no.:
    8.03977.0250

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    81 - 83 °C
  • Molar Mass:
    180.21 g/mol
  • Boiling Point:
    342 °C (1013 hPa)
  • Flash Point:
    163 °C
  • Density:
    0.9 g/cm3 (20 °C)
  • Ignition Point:
    608 °C

Specification Test Results

About this item

 :
 :180.21 g/mol
Boiling Pt:342 °C (1013 hPa)
Melting Pt:81…83 °C
Density:0.9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-630-7
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS07
 :GHS09
CAS Number:486-25-9
Formula:C₁₃H₈O
MDL Number:MFCD00001141