

Hydrochloric acid fuming 37%, EMSURE® for analysis (max. 0.001 ppm Hg), Supelco®
Catalog # EM1.13386.2500
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:2.5 L
- Cat. No.:EM1.13386.2500
- Supplier no.:1.13386.2500
Specifications
Specification Test Results
- Al (Aluminium):≤ 0.05 ppm
- As (Arsenic):≤ 0.01 ppm
- Assay (alkalimetric):≥ 37.0 %
- Ba (Barium):≤ 0.02 ppm
- Be (Beryllium):≤ 0.02 ppm
- Bi (Bismuth):≤ 0.1 ppm
- Bromide (Br):≤ 50 ppm
- Ca (Calcium):≤ 0.5 ppm
- Cd (Cadmium):≤ 0.01 ppm
- Co (Cobalt):≤ 0.01 ppm
- Color:≤ 10 Hazen
- Cr (Chromium):≤ 0.02 ppm
- Cu (Copper):≤ 0.02 ppm
- Fe (Iron):≤ 0.2 ppm
- Free chlorine (Cl):≤ 0.5 ppm
- Ge (Germanium):≤ 0.05 ppm
- Hg (Mercury):≤ 0.001 ppm
- K (Potassium):≤ 0.1 ppm
- Li (Lithium):≤ 0.01 ppm
- Mg (Magnesium):≤ 0.1 ppm
- Mn (Manganese):≤ 0.01 ppm
- Mo (Molybdenum):≤ 0.02 ppm
- Na (Sodium):≤ 0.5 ppm
- NH₄ (Ammonium):≤ 1 ppm
- Ni (Nickel):≤ 0.02 ppm
- Pb (Lead):≤ 0.02 ppm
- Phosphate (PO₄):≤ 0.5 ppm
- Residue on ignition (as SO₄):≤ 5 ppm
- Sb (Antimony):≤ 0.002 ppm
- Sr (Strontium):≤ 0.01 ppm
- Sulfate (SO₄):≤ 1 ppm
- Sulfite (SO₃):≤ 1 ppm
- Ti (Titanium):≤ 0.1 ppm
- Tl (Thallium):≤ 0.05 ppm
- V (Vanadium):≤ 0.01 ppm
- Zn (Zinc):≤ 0.05 ppm
- Zr (Zirconium):≤ 0.1 ppm
Specification Test Results
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
:M-Clarity™ quality level = MQ300
:36.46
EINECS:231-595-7
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324