



Hydroxylammonium sulfate ≥99%, GR analytical reagent, Supelco®
EM1.04617.0500
:
- Pack type:Plastic bottle
- Size:500 g
- Cat. No.:EM1.04617.0500
- Supplier no.:1.04617.0500
- Assay:≥ 99 %
- Chloride (Cl):≤ 0.001 %
- As (Arsenic):≤ 0.00005 %
- Cu (Copper):≤ 0.0005 %
- Fe (Iron):≤ 0.0005 %
- Hg (Mercury):≤ 0.000001 %
- Pb (Lead):≤ 0.0005 %
- Sulfated ash (residue on ignition):≤ 0.1 %
:
:164.14 g/mol
:13,04853
:170 °C
:1.1 g/cm³ (20 °C)
:233-118-8
:2865
:8,III
:M-Clarity™ quality level = MQ100
:GHS08
:GHS05
:GHS07
:GHS09
:10039-54-0
:H₈N₂O₆S
:MFCD00044869