Order Entry
United States
ContactUsLinkComponent
Mercury (II) oxide, red, extra pure, Supelco®
Mercury (II) oxide, red, extra pure, Supelco®
Catalog # EM1.04465.0100
Supplier:  Avantor
CAS Number:  
Mercury (II) oxide, red, extra pure, Supelco®
Catalog # EM1.04465.0100
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g
  • Environmentally Preferable:
  • Cat. No.:
    EM1.04465.0100
  • Supplier no.:
    1.04465.0100

Specifications

Specification Test Results

  • Solubility:
    0.052 g/l (25 °C)
  • Melting Point:
    >400 °C (decomposition)
  • Molar Mass:
    216.58 g/mol
  • Bulk Density:
    3000 kg/m3
  • Vapor Pressure:
    0.12 Pa (20 °C)
  • Density:
    11.1 g/cm3 (20 °C)
  • pH:
    6 - 7 (50 g/l, H2O, 20 °C) (slurry)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

About this item

 :
 :216.59 g/mol
EINECS:244-654-7
UN:1641
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:21908-53-2
Formula:HgO
MDL Number:MFCD00011045
Melting Pt:> 400 °C
Density:11 g/cm³ (20 °C)
ADR:6.1 II
Merck Index:13,05908