


1-Chloro-2,4-dinitrobenzene, GR for analysis, Supelco®
Catalog # EM1.02427.0025
Supplier: MilliporeSigma
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:25 g
- Cat. No.:EM1.02427.0025
- Supplier no.:1.02427.0025
Specifications
Specification Test Results
- Solubility:0.36 g/l (20 °C)
- Melting Point:48 - 52 °C
- Molar Mass:202.55 g/mol
- Bulk Density:760 kg/m3
- Boiling Point:315 °C (1013 hPa)
- Vapor Pressure:1 hPa (106 °C)
- Flash Point:194 °C
- Explosion Limit:1.9 - 22 %(V)
- Density:1.7 g/cm3 (20 °C)
- Ignition Point:464 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
About this item
:
:202.55 g/mol
Merck Index:13,02155
Boiling Pt:315 °C (1013 hPa)
Boiling Pt:315 °C (1013 hPa)
Melting Pt:52…54 °C
Melting Pt:52 - 54 °C
Density:1.7 g/cm³ (20 °C)
Flash Pt:194 °C
Flash Pt:194 °C
EINECS:202-551-4
UN:3441
ADR:6.1,II
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
CAS Number:97-00-7
Formula:C₆H₃ClN₂O₄
MDL Number:MFCD00007075