ContactUsLinkComponent
Temperature Indicator Strip, Therapak®
Temperature Indicator Strip, Therapak®
Catalog # 89171-046
Supplier:  Therapak, LLC
undefined
Temperature Indicator Strip, Therapak®
Catalog # 89171-046
Supplier:  Therapak, LLC
Supplier Number:  40006
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Temperature range:
    32...38 °C (90...100 °F)
  • Description:
    Temperature Indicator Strip
  • Cat. No.:
    89171-046
  • Supplier no.:
    40006
Specifications
About this item
Temperature Indicator Strips indicate the temperature of the specimen within minutes after the initial collection
As an integral part of the drug testing process, the temperature indicator strip confirms that the voided specimen was indeed within temperature expectations eliminating doubt at the point of collection that a specimen was subject to adulteration. Temperature readings can be viewed in both Celsius and Fahrenheit.