Order Entry
Puerto Rico
ContactUsLinkComponent
POWERSTAT with ACE Warning Glow, Ace Glass
POWERSTAT with ACE Warning Glow, Ace Glass
Catalog # 89076-322
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
POWERSTAT with ACE Warning Glow, Ace Glass
Catalog # 89076-322
Supplier:  Ace Glass
Supplier Number:  12081-05
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Output:
    0 - 120 V
  • Description:
    Powerstat® transformer with ACE warning glow
  • Cat. No.:
    89076-322
  • Electrical:
    120 V, 50/60 Hz, 10 A
  • Supplier no.:
    12081-05

Specifications

About this item

Warns of lethal shock hazard at low voltage settings

Light admonishes operator to correct the situation by reversing the plug in the line socket. The warning light also reappears when hazardous transformer output voltage is reached (50-55 V). When the output voltage reaches 50-55 V, sometimes enough to be lethal, but usually causing an unpleasant sticking sensation, the warning light appears. Supplied with three-wire neoprene power cord with NEMA plug, and instruction sheet.