Anti-SARS MATRIX Rabbit Polyclonal Antibody
89415-850
: Prosci
- Antibody Type:Primary
- Antigen Name:Membrane protein
- Antigen Symbol:M
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Size:0.1 mg
- Form:Liquid
- Gene ID:P59596
- Antigen Synonyms:M
- Storage Buffer:SARS Matrix Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:SARS Matrix antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:SARS matrix antibody was raised against a synthetic peptide corresponding to 15 amino acids at the carboxy terminus of the SARS matrix protein. The immunogen is located within the last 50 amino acids of SARS Matrix.
- Tested Applications:E
- Purification:SARS Matrix Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89415-850
- Supplier no.:3529
SARS Matrix Antibody: A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. The M protein (Membrane protein, Matrix protein) is one of the major structural viral proteins. It is an integral membrane protein involved in the budding of the viral particles and interacts with S (Spike) protein and the nucleocapsid protein.
Type: Primary
Antigen: M
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus