Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-SARS SPIKE Rabbit Polyclonal Antibody
Anti-SARS SPIKE Rabbit Polyclonal Antibody
Catalog # 89415-636
Supplier:  Prosci
undefined
Anti-SARS SPIKE Rabbit Polyclonal Antibody
Catalog # 89415-636
Supplier:  Prosci
Supplier Number:  3221

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Spike glycoprotein
  • Antigen Symbol:
    S
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    P59594
  • Antigen Synonyms:
    S
  • Storage Buffer:
    SARS Spike Antibody is supplied in PBS containing 0.02% sodium azide.
  • Storage Temperature:
    SARS Spike antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    SARS Spike antibody was raised against a synthetic peptide corresponding to amino acids near the center of the SARS Spike glycoprotein. The immunogen is located within amino acids 410 - 460 of SARS Spike.
  • Tested Applications:
    E
  • Purification:
    SARS Spike Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    89415-636
  • Supplier no.:
    3221

Specifications

About this item

SARS Spike Antibody: A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. SARS infection can be mediated by the binding of the viral spike protein, a glycosylated 139 kDa protein and the major surface antigen of the virus, to the angiotensin-converting enzyme 2 (ACE2) on target cells. This binding can be blocked by a soluble form of ACE2.

Type: Primary
Antigen: S
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus