Specifications
- Antibody Type:Primary
- Antigen Name:phorbol-12-myristate-13-acetate-induced protein 1
- Antigen Symbol:Pmaip1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q9JM54
- Antigen Synonyms:Pmaip1
- Storage Buffer:Noxa Antibody is supplied in PBS containing 0.02% sodium azide.
- Storage Temperature:Noxa antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Noxa antibody was raised against a synthetic peptide corresponding to 17 amino acids at the amino terminus of mouse Noxa. The immunogen is located within the first 50 amino acids of Noxa.
- Tested Applications:E, WB, IHC, IF
- Purification:Noxa Antibody is immunoaffinity chromatography purified IgG.
- Cat. No.:89415-474
- Supplier no.:2437
Specifications
About this item
Noxa Antibody: Apoptosis is related to many diseases and development. The p53 tumor-suppressor protein induces apoptosis through transcriptional activation of several genes including p53R2, p53AIP1, and PUMA. A new p53 target gene, Noxa, was recently identified, which encodes a protein belonging to the subfamily of BH3-only proapoptic proteins. Noxa and PUMA are both transcriptional targets of p53 and BH3-only proteins. X-ray irradiation increased p53-dependent Noxa mRNA and protein levels. Noxa, when ectopically expressed, interacted with anti-apoptotic Bcl-2 family members, resulting in the activation of caspase-9. Noxa, like PUMA, localized to mitochondria and induces apoptosis in response to p53. Noxa and PUMA may represent direct mediators of p53-induced apoptosis. Increased levels of p53 and its target gene Noxa was found in the impaired tumor development.
Type: Primary
Antigen: Pmaip1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat