- Antibody Type:Primary
- Antigen Name:tumor necrosis factor (ligand) superfamily, member 10
- Antigen Symbol:TNFSF10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P50591
- Antigen Synonyms:TNFSF10
- Storage Buffer:Trail Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 32 kDa Observed: 35 kDa
- Storage Temperature:Trail antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:TRAIL antibody was raised against a peptide corresponding to 17 amino acids near the carboxy terminus of human TRAIL. The immunogen is located within the last 50 amino acids of Trail.
- Tested Applications:E, WB, IHC
- Purification:Trail Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89415-216
- Supplier no.:1113
Trail Antibody: Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF and Fas ligand in the TNF family through their death domain containing receptors, TNFR1 and Fas. A novel member in the TNF family was recently identified and designated TRAIL (for TNF-related apoptosis-inducing ligand) and Apo-2L (for Apo-2 ligand)1, TRAIL is a type II membrane protein and expressed in a variety of human tissues. Two novel death domain containing receptors DR4 and DR5 have been identified as the receptor for TRAIL3-6. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kappa B activation in many tissues and cells.
Type: Primary
Antigen: TNFSF10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human