ContactUsLinkComponent
Tris-Buffered Saline with Tween Solution (10X), Genetex
Tris-Buffered Saline with Tween Solution (10X), Genetex
Catalog # 89306-978
Supplier:  Genetex
CAS Number:  
Tris-Buffered Saline with Tween Solution (10X), Genetex
Catalog # 89306-978
Supplier:  Genetex
Supplier Number:  GTX30976
CAS Number:  
Specifications
  • Description:
    Tris-Buffered Saline (10X)
  • Cat. No.:
    89306-978
  • Supplier no.:
    GTX30976
Specifications
About this item
Dilute this 10X buffer 10 times, mix well, and store 1 X buffer solution at room temperature or 2-8oC
Liquid form. Store at room temperature. If stored at 2°C-8°C, some sodium chloride will be precipitated, if this occurs, bring the buffer to room temperature and mix well till all sodium chloride is dissolved.