Order Entry
Puerto Rico
ContactUsLinkComponent
 
SP Bel-Art Microcentrifuge Floating Rack, Bel-Art Products, a part of SP
  20901-905
undefined
SP Bel-Art Microcentrifuge Floating Rack, Bel-Art Products, a part of SP
  20901-905
 :  F18876-0000

 

  • Hole Ø:
    10.8 mm (⁷/₁₆")
  • Description:
    MCT Floating Rack
  • Dimensions:
    119 x 78 mm (4¹¹⁄₁₆ x 3¹⁄₁₆")
  • Cat. No.:
    20901-905
  • Supplier no.:
    188760000

 

 

Incubate up to 24 microcentrifuge tubes in a water bath (choose from 1.5 mL or 0.5 mL size).

  • Floating rack design
  • Easy to load
  • Snug fit for secure holding of microtubes

Easy to load, the microtubes are held snugly and then released when the rack is pressed down on a benchtop.