Order Entry
Puerto Rico
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
Catalog # 89139-054
Supplier:  Biotium
CAS Number:  
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
Catalog # 89139-054
Supplier:  Biotium
Supplier Number:  40011
CAS Number:  

Specifications

  • Size:
    10 mg
  • Cat. No.:
    89139-054
  • Supplier no.:
    40011

Specifications

About this item

 :
 :350.25 g/mol
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681