ContactUsLinkComponent
Quick-Prep™ Urinalysis Kit, Globe Scientific
Quick-Prep™ Urinalysis Kit, Globe Scientific
  89497-594
 :  
undefined
Quick-Prep™ Urinalysis Kit, Globe Scientific
  89497-594
 :  3825
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Pack type:
    1000 Tubes And Pipettes Per Case
  • Description:
    Quick-Prep™ Urinalysis Kit
  • Cat. No.:
    89497-594
  • Supplier no.:
    3825
The Quick-Prep™ Urinalysis Kit is a tube and pipet system that isolates 1mL of urine sediment for microscopic examination.
The kit includes the specially designed transfer pipet and 12mL urine centrifuge tube. This system is used to isolate 1mL of urine sediment stain and allow easy pour off of the balance.
Quick-Prep™ has been designed for use with the 10 Chamber Urinalysis Slide (see 89206-938 series).