ContactUsLinkComponent
Green Supreme HEPA Vac, Atrix
  101640-734
cUL Listed
UL Listed
undefined
Green Supreme HEPA Vac, Atrix
  101640-734
 :  VACGRNS
cUL Listed
UL Listed
  • Capacity:
    1 gal.
  • Color:
    Green
  • Description:
    Green Supreme HEPA Vacuum with 1,000 Hour Motor
  • Cat. No.:
    101640-734
  • Supplier no.:
    VACGRNS
The Green Supreme 1 Gallon HEPA dry particulate vacuum is ideal for the RRP Contractor
Replacement Filters are designed to capture fumes, atmospheric dust, black toner and all color toner and other ultra-fine particulate. Compatible with 3M vacuums.
With its self contained HEPA filter, hose plugs and filter plugs, the VACGRNS makes filter changes, transporting and storing safe. Vacuum is compact and portable. HEPA Efficiency: 99.97% @ .3 microns.
 : Includes self-storing utensils (cover insert), 6' ESD Safe hose, two hose plugs, 16" ESD gooseneck, two 18" wands, utility nozzle with brush, shoulder strap with D-rings, HEPA filter, 10' power cord. Not For Use With Asbestos