Battery Backpack Vac, Atrix
Catalog # 100720-986
Supplier: Atrix International
Specifications
- Capacity:8 Qt.
- Color:Black
- Description:Battery backpack vacuum with 4-stage filtration
- Environmentally Preferable:
- Current:12 A
- Power:1400 W
- Voltage:36 V
- Cat. No.:100720-986
- Supplier no.:VACBP36V
Specifications
About this item
The Atrix Battery Backpack is perfect for cleaning confined areas and where corded electric vacuums are not a viable option
The Atrix International VACBP6-5P HEPA Replacement Filter Bags are designed to capture particulates as small as 0.3 microns when using the Backpack Series Vacuum Systems.
The battery feature eliminates all safety hazards that corded equipment may cause. Users are granted the freedom to clean anywhere such as on scaffolding, entry ways, attics, or in stairwells. Ergonomic and comfortable fit for all sizes. The VACBP36V has a continuous run time of 50-55 minutes and features a convenient on/off switch which allows users to power down between work stations and extend battery life.
Ordering information: Includes 36V flow-through motor, hose, adjustable extension wand, hard floor tool, turbo brush, oval brush, furniture brush, hose and filter plugs, adapter, gooseneck tool, 36V Battery, Battery Harness and Charger. Cannot Ship Air Freight.