Order Entry
Puerto Rico
ContactUsLinkComponent
Diehls-Alder Pressure Tube, Ace Glass Incorporated
Diehls-Alder Pressure Tube, Ace Glass Incorporated
Catalog # 89063-438
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Diehls-Alder Pressure Tube, Ace Glass Incorporated
Catalog # 89063-438
Supplier:  Ace Glass
Supplier Number:  8651-10
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Ext.Ø:
    38 mm
  • Capacity:
    100 ml
  • Description:
    Diehls-Alder Pressure Tube
  • Length:
    178 mm
  • Cat. No.:
    89063-438
  • Supplier no.:
    8651-10

Specifications

About this item

Heavy wall with lip for crown cap closure

Used for polymerization and analysis of halogens by thermal decomposition in the presence of lime.