Order Entry
United States
ContactUsLinkComponent
Labjaws Lab Frame Set, Extra Large, Ace Glass
Labjaws Lab Frame Set, Extra Large, Ace Glass
Catalog # 89084-708
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Labjaws Lab Frame Set, Extra Large, Ace Glass
Catalog # 89084-708
Supplier:  Ace Glass
Supplier Number:  11165-05
Product available on GSA Advantage®
Small Business Enterprise

Specifications

  • Description:
    Labjaws Lab Frame Set, Extra Large, 122 x 183 cm
  • Environmentally Preferable:
  • Cat. No.:
    89084-708
  • Supplier no.:
    11165-05

Specifications

About this item

This wall-type Lab Frame Set can be built into assemblies as large as 1.2 x 1.8 meters

Frame consists of (10) 5 cm rods, (3) 46 cm rods, (2) 91 cm rods, (10) 122 cm rods, (38) S connectors, (4) rod end connectors and (6) feet.

Can be used vertically or horizontally. Includes six 31 cm rods and feet for securing frame to a wall. Apparatus is mountable on either side of frame.