LabJaws Clamp, Talboys by Troemner, Ace Glass
Catalog # 89083-254
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:LabJaws Clamp Extension, 2 Prong, 6.5 cm Medium Grip
- Cat. No.:89083-254
- Arm length:127 mm (5")
- Supplier no.:11070-26
Specifications
About this item
These Labjaw Clamps are 2 prong extension clamps with a single adjustment screw for applications where minimal jaw adjustment is required
Vinylized jaws.