Order Entry
Puerto Rico
ContactUsLinkComponent
 
Labjaws Lab Frame Set, Large, Ace Glass
  89084-706
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Labjaws Lab Frame Set, Large, Ace Glass
  89084-706
 :  Ace Glass
 :  11163-07
Product available on GSA Advantage®
Small Business Enterprise

 

  • Description:
    Labjaws Lab Frame Set, Large, 122 x 122 cm
  • Cat. No.:
    89084-706
  • Supplier no.:
    11163-07

 

 

Elaborate apparatus setups are possible with this set

Frame consists of (10) 5 cm rods, (3) 46 cm rods, (2) 91 cm rods, (10) 122 cm rods, (38) S connectors, (4) rod end connectors and (6) feet.

It can be assembled into a support as large as 122 cm square with a 46 cm base.