ContactUsLinkComponent
Labjaws Frame Connectors, Ace Glass
Catalog # 89084-742
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Labjaws Frame Connectors, Ace Glass
Catalog # 89084-742
Supplier:  Ace Glass
Supplier Number:  11168-19
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Labjaws Frame Connectors, 90° Angle
  • Environmentally Preferable:
  • Cat. No.:
    89084-742
  • Supplier no.:
    11168-19
Specifications
About this item
Holds rods firmly at a 90° angle
Adjust by tightening square-headed screws. Use wrench or a screwdriver. Holes carefully sized to eliminate play.