Labjaws Lab Frame Set, Medium, Ace Glass
Catalog # 89084-710
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Specifications
- Description:Labjaws Lab Frame Set, Medium, 61 x 122cm
- Environmentally Preferable:
- Cat. No.:89084-710
- Supplier no.:11162-08
Specifications
About this item
This set can be assembled into a support for medium sized labs in either a horizontal or vertical position
Frame consists of (8) 5 cm rods, (2) 46 cm rods, (5) 61 cm rods, (2) 91 cm rods, (4) 122 cm rods, (24) S connectors, (4) rod end connectors and (4) feet.
The base rods are 46 cm long.