ContactUsLinkComponent
Labjaws End-To-End Rod Connectors, Ace Glass
Labjaws End-To-End Rod Connectors, Ace Glass
Catalog # 89084-714
Supplier:  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
Labjaws End-To-End Rod Connectors, Ace Glass
Catalog # 89084-714
Supplier:  Ace Glass
Supplier Number:  11175-23
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Description:
    Labjaws End-To-End Rod Connectors, Alloy
  • Dimensions:
    22×51 mm
  • Cat. No.:
    89084-714
  • Supplier no.:
    11175-23
Specifications
About this item
These strong alloy end-to-end connectors permit end-to-end joining of rods
Precision boring of connector ensures perfect aligning of rods. Corrosion resistant. Provided with two set screws.