ContactUsLinkComponent
MicroBlast™ Canned Air, MicroCare
MicroBlast™ Canned Air, MicroCare
Catalog # 46620-508
Supplier:  MicroCare
Small Business Enterprise
undefined
MicroBlast™ Canned Air, MicroCare
Catalog # 46620-508
Supplier:  MicroCare
Supplier Number:  MCC-DST
Small Business Enterprise
Specifications
  • Volume:
    284 g (10 oz.)
  • Description:
    MicroBlast™ Canned Air
  • Cat. No.:
    46620-508
  • Supplier no.:
    MCC-DST
Specifications
About this item
Ozone-safe dry circuit cleaner
High pressure trigger sprayer uses 70psi to blow away contamination. Micro-filtered to <0.2u. Cleans keyboards, disk drives, and PCBs. Nontoxic, nonflammable, plastic-safe. No residues. Non-conductive.