Specifications
- Pack type:Plastic ampoule
- Description:PBS Tablets
- Size:10 Tablets
- Cat. No.:80058-556
- Supplier no.:524650-1EA
Specifications
About this item
Dissolving one tablet in 1 liter of deionized H₂O yields 140 mM NaCl, 10 mM phosphate buffer, and 3 mM KCl, pH 7.4 at 25°C.