Di-tert-butyl pyrocarbonate ≥99%, crystals
Supplier: Chem Impex International
New Product
Certificates
About this item
CAS Number:24424-99-5
:
Formula:C₁₀H₁₈O₅
MDL Number:MFCD00008805
:218.25 g/mol
UN:2930
ADR:6.1,I
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Molecular Formula:C10H18O5
- Molecular Weight:218.25
- Melting Point:20-25 °C
- Boiling Point:56-57°C/0.5mmHg
- MDL Number:MFCD00008805
- Storage Temperature:Store at 0-8°C
- Flash Point:37 ºC
- Refractive Index:1.409
- Density:0.95 g/mL
- Color:Colorless or White
Specification Test Results
Related Information
Certificates
Search by Lot NumberAT-00128.pdf
Datasheet
VWR GHS Compliance Letter (US)