SP Bel-Art Book Bottle, Bel-Art Products, a part of SP
Catalog # 16126-612
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacity:5 L (1.3 gal.)
- Description:Book Bottle
- Dimensions:8.9 x 24.1 x 37.5 cm (3⁷/₁₆ x 9½ x 14¾")
- Environmentally Preferable:
- Cat. No.:16126-612
- Supplier no.:109390005
Specifications
About this item
Unique, polypropylene book bottle stacks book-like on shelves for efficient use of space.
- Polypropylene construction
- Easy to stack
- Effective, efficient use of space
- Convenient handle and grip hold for easy removal from shelves
In addition to the top handle, a convenient grip hold in the front facilitates removal from a shelf.
Graduated markings on front in liters.