Order Entry
Puerto Rico
ContactUsLinkComponent
Activate™ Bleach Dilution System, Deardorff Fitzsimmons
Activate™ Bleach Dilution System, Deardorff Fitzsimmons
Catalog # 89136-482
Activate™ Bleach Dilution System, Deardorff Fitzsimmons
Catalog # 89136-482
Supplier Number:  77062

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    89136-482
  • Concentration:
    5.25%
  • Supplier no.:
    77062

Specifications

About this item

The innovative Activate™ Bleach Dilution System dispenses the 10% bleach solution widely recommended by CDC and other nationally recognized authorities

This two-bottle fixed-ratio trigger sprayer automatically mixes and dilutes 5.25% bleach concentrate with tap water to make a 5000 ppm bleach disinfectant solution for use on hard, non-porous surfaces. The sprayer keeps the bleach separate from the water until you spray, keeping the 10% solution fresh and effective. Refill the water cartridge with tap water and lock in the disposable bleach cartridges and system is ready for use.

Its fast contact times, sporicidal action and innovative dispenser make it the most practical way to use bleach in the healthcare and laboratory environment.

Ordering information: Includes Six, 11 oz. Bleach Cartridges, Two 5000 ppm spayer heads, and Two 11 oz. Water Bottles.