ContactUsLinkComponent
Simple Machines, Simple Wedge Model
Simple Machines, Simple Wedge Model
Catalog # 470210-814
Supplier:  Wards
undefined
Simple Machines, Simple Wedge Model
Catalog # 470210-814
Supplier:  Wards
Supplier Number:  WDMS14
Specifications
  • Description:
    Simple Machines, Simple Wedge Model
  • Cat. No.:
    470210-814
  • Supplier no.:
    WDMS14
Specifications
About this item
This wooden machine is elegantly designed to demonstrate how a wedge can increase mechanical advantage.
  • Demonstrate mechanical advantages
  • Includes manual
Easy for students to use and measure. Includes an instruction manual to facilitate student and teacher use of the apparatus.