Order Entry
Puerto Rico
ContactUsLinkComponent
India ink solution
India ink solution
Catalog # 89428-916
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
India ink solution
Catalog # 89428-916
Supplier:  Hardy Diagnostics
Supplier Number:  Z64
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Pack type:
    Dropping vial
  • Size:
    15 ml
  • Cat. No.:
    89428-916
  • Supplier no.:
    Z64

Specifications

About this item

 :
CAS Number:8046-52-4
MDL Number:MFCD00147244