Phosphate Buffered Saline (PBS), Hardy Diagnostics
Catalog # 89498-948
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Bottle
- Description:PH 7.5
- Size:125ml
- Storage Temperature:2-8C
- Cat. No.:89498-948
- Fill volume:100 ml
- Supplier no.:U137
Specifications
About this item
Phosphate Buffered Saline (PBS), Ph 7.5, 100ml fill, 125ml polypropylene bottle, order by the each, by Hardy Diagnostics
Hardy Diagnostics Phosphate Buffered Saline (pH of 7.5) is recommended as a diluent for quantitative culturing or as a rinsing agent in microbiological, immunological, histological and molecular testing procedures.