R2A Agar, Hardy Diagnostics
52428-870
Product available on GSA Advantage®
:
- Pack type:Tube
- Description:Agar
- Size:20 x 125 mm
- Storage Temperature:2-30C
- Cat. No.:52428-870
- Fill volume:18 ml
- Supplier no.:Q77
R2A Agar, 18ml fill, 20x125mm tube deep, order by the package of 20, by Hardy Diagnostics.