Order Entry
Puerto Rico
ContactUsLinkComponent
 
Peptone, Hardy Diagnostics
Catalog # 89423-614
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
Peptone, Hardy Diagnostics
Catalog # 89423-614
Supplier:  Hardy Diagnostics
Supplier Number:  LP037B
Product available on GSA Advantage®
CAS Number:  

Specifications

  • Description:
    Bateriological
  • Size:
    500 g
  • Storage Temperature:
    15-30C
  • Cat. No.:
    89423-614
  • Supplier no.:
    LP037B

Specifications

About this item

Peptone, Bacteriological, 500gm, by Oxoid